missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine 98.0+%, TCI America™

Catalog No. B50111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50111G 1 g
B50115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50111G Supplier TCI America Supplier No. B50111G
Only null left

Chemical Identifiers

CAS 85466-66-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00151890
InChI Key AJGJINVEYVTDNH-UHFFFAOYNA-N
Synonym boc-n-me-d-phe-oh, boc-d-mephe-oh, n-boc-n-methyl-d-phenylalanine, n-tert-butoxycarbonyl-n-methyl-d-phenylalanine, n-methyl-boc-d-phenylalanine, r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid, 2r-2-n-tert-butoxycarbonyl-n-methylamino-3-phenylpropionic acid, boc-d-me-phe-oh, ambotzbaa1044, 2r-2-tert-butoxycarbonyl methyl amino-3-phenylpropanoic acid
PubChem CID 7015127
IUPAC Name 2-{[(tert-butoxy)carbonyl](methyl)amino}-3-phenylpropanoic acid
SMILES CN(C(CC1=CC=CC=C1)C(O)=O)C(=O)OC(C)(C)C

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 1 g
Formula Weight 279.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-N-methyl-D-phenylalanine