missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-tyrosine Methyl Ester 98.0+%, TCI America™

Catalog No. B20055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20055G 5 g
B200525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20055G Supplier TCI America Supplier No. B20055G

Chemical Identifiers

CAS 4326-36-7
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.34
MDL Number MFCD00191181
InChI Key NQIFXJSLCUJHBB-STGVRZAANA-N
Synonym boc-tyr-ome, boc-l-tyrosine methyl ester, n-boc-l-tyrosine methyl ester, n-tert-butoxycarbonyl-l-tyrosine methyl ester, methyl n-boc-l-tyrosinate, methyl 2s-2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, methyl 2s-2-tert-butoxy carbonyl amino-3-4-hydroxyphenyl propanoate, s-methyl 2-tert-butoxycarbonyl amino-3-4-hydroxyphenyl propanoate, s-2-tert-butoxycarbonylamino-3-4-hydroxy-phenyl-propionic acid methyl ester
PubChem CID 7019130
IUPAC Name methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 106°C
Color White
Quantity 5 g
Formula Weight 295.34
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-tyrosine Methyl Ester