missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-tyrosine 98.0+%, TCI America™

Catalog No. B16405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16405G 5 g
B164025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16405G Supplier TCI America Supplier No. B16405G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3978-80-1
Molecular Formula C14H19NO5
Molecular Weight (g/mol) 281.31
MDL Number MFCD00037179
InChI Key CNBUSIJNWNXLQQ-LDGXTIHJNA-N
Synonym boc-tyr-oh, boc-l-tyrosine, n-boc-l-tyrosine, n-tert-butoxycarbonyl-l-tyrosine, n-boc-tyrosine, n-t-boc-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl, chembl65386, n-t-butoxycarbonyl-l-tyrosine, n-tert-butoxycarbonyl tyrosine
PubChem CID 117439
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 281.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-tyrosine