missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-tert-leucine 98.0+%, TCI America™

Catalog No. p-7123146
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37545G 5 g
B375425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37545G Supplier TCI America Supplier No. B37545G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 62965-35-9
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.29
MDL Number MFCD00065574
InChI Key LRFZIPCTFBPFLX-JLDDOWRYNA-N
Synonym n-boc-l-tert-leucine, boc-tle-oh, boc-l-tert-leucine, s-n-tert-butoxycarbonyl-tert-leucine, s-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-l-alpha-t-butylglycine, boc-alpha-t-butylglycine, boc-tbu-gly-oh, boc-l-tle-oh, n-tert-butoxycarbonyl-l-tert-leucine
PubChem CID 2734668
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](C(O)=O)C(C)(C)C

Specifications

Melting Point 119°C
Color White
Quantity 5 g
Formula Weight 231.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-tert-leucine