missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-pyroglutamic Acid 98.0+%, TCI America™
Supplier: TCI America B408325G
Specifications
| N-(tert-Butoxycarbonyl)-L-pyroglutamic Acid | |
| 119°C | |
| C10H15NO5 | |
| 25 g | |
| MJLQPFJGZTYCMH-LURJTMIESA-N | |
| (2S)-1-[(tert-butoxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid | |
| 104412 | |
| ≥98.0% (HPLC,T) |
| 53100-44-0 | |
| White | |
| MFCD00672316 | |
| boc-pyr-oh, n-boc-l-pyroglutamic acid, s-boc-5-oxopyrrolidine-2-carboxylic acid, boc-l-pyroglutamic acid, 2s-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylic acid, 5-oxo-, 1-1,1-dimethylethyl ester, 2s, 1-tert-butoxycarbonyl-5-oxo-l-proline, bocpyr, 2s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid | |
| CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(O)=O | |
| 229.23 | |
| 229.23 | |
| Crystalline Powder |
Chemical Identifiers
| 53100-44-0 | |
| 229.23 | |
| MJLQPFJGZTYCMH-LURJTMIESA-N | |
| 104412 | |
| CC(C)(C)OC(=O)N1[C@@H](CCC1=O)C(O)=O |
| C10H15NO5 | |
| MFCD00672316 | |
| boc-pyr-oh, n-boc-l-pyroglutamic acid, s-boc-5-oxopyrrolidine-2-carboxylic acid, boc-l-pyroglutamic acid, 2s-1-tert-butoxy carbonyl-5-oxopyrrolidine-2-carboxylic acid, s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylic acid, 5-oxo-, 1-1,1-dimethylethyl ester, 2s, 1-tert-butoxycarbonyl-5-oxo-l-proline, bocpyr, 2s-1-tert-butoxycarbonyl-5-oxopyrrolidine-2-carboxylic acid | |
| (2S)-1-[(tert-butoxy)carbonyl]-5-oxopyrrolidine-2-carboxylic acid |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator