missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-prolinol 98.0+%, TCI America™

Catalog No. p-7119934
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30775G 5 g
B307725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30775G Supplier TCI America Supplier No. B30775G
Only null left

Chemical Identifiers

CAS 69610-40-8
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD00066232
InChI Key BFFLLBPMZCIGRM-QMMMGPOBSA-N
Synonym n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
PubChem CID 643448
IUPAC Name tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-prolinol