missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-phenylalanine 99.0+%, TCI America™

Catalog No. p-7118998
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B13325G 5 g
B133225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B13325G Supplier TCI America Supplier No. B13325G
Only null left

Chemical Identifiers

CAS 13734-34-4
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.31
MDL Number MFCD00002663
InChI Key ZYJPUMXJBDHSIF-NSHDSACASA-N
Synonym boc-phe-oh, boc-l-phenylalanine, boc-phenylalanine, n-tert-butoxycarbonyl-l-phenylalanine, n-boc-l-phenylalanine, n-t-boc-l-phenylalanine, n-alpha-t-boc-l-phenylalanine, n-t-butyloxycarbonyl-l-phenylalanine, l-phenylalanine, n-1,1-dimethylethoxy carbonyl, boc-phe
PubChem CID 77857
IUPAC Name (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 5 g
Formula Weight 265.31
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-phenylalanine