missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-phenylalanine Methyl Ester 98.0+%, TCI America™

Catalog No. p-7123435
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B42595G 5 g
B425925G 25 g
2 options

Catalog No. B42595G

Supplier: TCI America B42595G

Only null left

Chemical Identifiers

CAS 51987-73-6
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.34
MDL Number MFCD00076977
InChI Key SDSWSVBXRBXPRL-UHFFFAOYNA-N
Synonym boc-phe-ome, boc-l-phenylalanine methyl ester, l-boc-phenyl-alanine methyl ester, n-boc-l-phenylalanine methyl ester, n-tert-butoxycarbonyl-l-phenylalanine methyl ester, s-methyl 2-tert-butoxycarbonyl amino-3-phenylpropanoate, s-2-tert-butoxycarbonylamino-3-phenyl-propionic acid methyl ester, methyl 2s-2-tert-butoxycarbonyl amino-3-phenylpropanoate, t-boc-l-phe-ome, pubchem13507
PubChem CID 9882138
IUPAC Name methyl 2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
SMILES COC(=O)C(CC1=CC=CC=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 39°C
Color White
Quantity 5 g
Formula Weight 279.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-phenylalanine Methyl Ester
EINECSNumber (3)-4367
TSCA No