missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-homophenylalanine 98.0+%, TCI America™
Supplier: TCI America B43565G
| Quantity | 5 g |
|---|
Chemical Identifiers
| 100564-78-1 | |
| 279.336 | |
| MCODLPJUFHPVQP-LBPRGKRZSA-N | |
| 7018726 | |
| CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O |
| C15H21NO4 | |
| MFCD00076904 | |
| boc-l-homophenylalanine, boc-homophe-oh, boc-hophe-oh, boc-l-homo-phe, s-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid, boc-homophenylalanine, boc-hfe-oh, n-boc-l-homophenylalanine, n-alpha-boc-l-homophenylalanine | |
| (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid |
Specifications
| 78°C | |
| 100564-78-1 | |
| C15H21NO4 | |
| boc-l-homophenylalanine, boc-homophe-oh, boc-hophe-oh, boc-l-homo-phe, s-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid, boc-homophenylalanine, boc-hfe-oh, n-boc-l-homophenylalanine, n-alpha-boc-l-homophenylalanine | |
| CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O | |
| 5 g | |
| 7018726 | |
| ≥98.0% (HPLC,T) |
| N-(tert-Butoxycarbonyl)-L-homophenylalanine | |
| White | |
| MFCD00076904 | |
| MCODLPJUFHPVQP-LBPRGKRZSA-N | |
| (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid | |
| 279.336 | |
| 279.34 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator