missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-homophenylalanine 98.0+%, TCI America™

Catalog No. B43561G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43561G 1 g
B43565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43561G Supplier TCI America Supplier No. B43561G

Chemical Identifiers

CAS 100564-78-1
Molecular Formula C15H21NO4
Molecular Weight (g/mol) 279.336
MDL Number MFCD00076904
InChI Key MCODLPJUFHPVQP-LBPRGKRZSA-N
Synonym boc-l-homophenylalanine, boc-homophe-oh, boc-hophe-oh, boc-l-homo-phe, s-2-tert-butoxycarbonyl amino-4-phenylbutanoic acid, boc-homophenylalanine, boc-hfe-oh, n-boc-l-homophenylalanine, n-alpha-boc-l-homophenylalanine
PubChem CID 7018726
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-phenylbutanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(=O)O

Specifications

Melting Point 78°C
Color White
Quantity 1 g
Formula Weight 279.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-homophenylalanine
TSCA No
Recommended Storage Refrigerator