missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-aspartic Acid 98.0+%, TCI America™

Catalog No. B22705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B22705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B22705G Supplier TCI America Supplier No. B22705G

Chemical Identifiers

CAS 13726-67-5
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00037279
InChI Key KAJBMCZQVSQJDE-YFKPBYRVSA-N
Synonym boc-asp-oh, n-tert-butoxycarbonyl-l-aspartic acid, boc-l-aspartic acid, boc-l-asp-oh, n-boc-l-aspartic acid, boc-aspartic acid, boc-l-aspartic acid-oh, n-tert-butyloxycarbonyl-l-aspartic acid, n-t-butoxycarbonyl-l-aspatic acid, 2s-2-tert-butoxycarbonyl amino butanedioic acid
PubChem CID 99718
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 233.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-aspartic Acid