missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-alaninol 98.0+%, TCI America™

Catalog No. B19535G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19535G 5 g
B19531G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19535G Supplier TCI America Supplier No. B19535G

Chemical Identifiers

CAS 79069-13-9
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00043121
InChI Key PDAFIZPRSXHMCO-LURJTMIESA-N
Synonym boc-l-alaninol, n-boc-l-alaninol, boc-alaninol, s-2-boc-amino-1-propanol, n-tert-butoxycarbonyl-l-alaninol, s-tert-butyl 1-hydroxypropan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1-propanol, tert-butyl n-2s-1-hydroxypropan-2-yl carbamate, s---2-tert-butoxycarbonylamino-1-propanol
PubChem CID 7023103
IUPAC Name tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
SMILES C[C@@H](CO)NC(=O)OC(C)(C)C

Specifications

Melting Point 60°C
Color White
Quantity 5 g
Formula Weight 175.23
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-alaninol