missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-alaninol 98.0+%, TCI America™

Supplier:  TCI America B19531G

Quantity 1 g

 View more versions of this product

Catalog No. B19531G


Only null left

Description

Description

Specifications

Chemical Identifiers

79069-13-9
175.23
PDAFIZPRSXHMCO-LURJTMIESA-N
7023103
C[C@@H](CO)NC(=O)OC(C)(C)C
C8H17NO3
MFCD00043121
boc-l-alaninol, n-boc-l-alaninol, boc-alaninol, s-2-boc-amino-1-propanol, n-tert-butoxycarbonyl-l-alaninol, s-tert-butyl 1-hydroxypropan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1-propanol, tert-butyl n-2s-1-hydroxypropan-2-yl carbamate, s---2-tert-butoxycarbonylamino-1-propanol
tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate

Specifications

60°C
79069-13-9
C8H17NO3
boc-l-alaninol, n-boc-l-alaninol, boc-alaninol, s-2-boc-amino-1-propanol, n-tert-butoxycarbonyl-l-alaninol, s-tert-butyl 1-hydroxypropan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1-propanol, tert-butyl n-2s-1-hydroxypropan-2-yl carbamate, s---2-tert-butoxycarbonylamino-1-propanol
C[C@@H](CO)NC(=O)OC(C)(C)C
175.23
7023103
≥98.0% (GC,N)
N-(tert-Butoxycarbonyl)-L-alaninol
White
MFCD00043121
PDAFIZPRSXHMCO-LURJTMIESA-N
tert-butyl N-[(2S)-1-hydroxypropan-2-yl]carbamate
1 g
175.23
Crystalline Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions