missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-L-2-phenylglycinol 98.0+%, TCI America™
$57.79 - $164.03
Chemical Identifiers
| CAS | 117049-14-6 |
|---|---|
| Molecular Formula | C13H19NO3 |
| Molecular Weight (g/mol) | 237.30 |
| MDL Number | MFCD00274206 |
| InChI Key | IBDIOGYTZBKRGI-UHFFFAOYNA-N |
| Synonym | s-tert-butyl 2-hydroxy-1-phenylethyl carbamate, boc-l-phenylglycinol, s-+-2-boc-amino-2-phenylethanol, n-boc-l-alpha-phenylglycinol, s-n-tert-butoxycarbonyl-2-phenylglycinol, s-2-tert-butoxycarbonylamino-2-phenylethanol, boc-phg-ol, n-boc-l-2-phenylglycinol, tert-butyl n-1s-2-hydroxy-1-phenylethyl carbamate, tert-butyl 1s-2-hydroxy-1-phenylethyl carbamate |
| PubChem CID | 7016462 |
| IUPAC Name | tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1 |
Chemical Identifiers
| 117049-14-6 | |
| 237.30 | |
| IBDIOGYTZBKRGI-UHFFFAOYNA-N | |
| 7016462 | |
| CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1 |
| C13H19NO3 | |
| MFCD00274206 | |
| s-tert-butyl 2-hydroxy-1-phenylethyl carbamate, boc-l-phenylglycinol, s-+-2-boc-amino-2-phenylethanol, n-boc-l-alpha-phenylglycinol, s-n-tert-butoxycarbonyl-2-phenylglycinol, s-2-tert-butoxycarbonylamino-2-phenylethanol, boc-phg-ol, n-boc-l-2-phenylglycinol, tert-butyl n-1s-2-hydroxy-1-phenylethyl carbamate, tert-butyl 1s-2-hydroxy-1-phenylethyl carbamate | |
| tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate |
Specifications
| 136°C | |
| White-Yellow | |
| MFCD00274206 | |
| IBDIOGYTZBKRGI-UHFFFAOYNA-N | |
| tert-butyl N-(2-hydroxy-1-phenylethyl)carbamate | |
| 237.30 | |
| 237.30 | |
| Crystalline Powder |
| 117049-14-6 | |
| C13H19NO3 | |
| s-tert-butyl 2-hydroxy-1-phenylethyl carbamate, boc-l-phenylglycinol, s-+-2-boc-amino-2-phenylethanol, n-boc-l-alpha-phenylglycinol, s-n-tert-butoxycarbonyl-2-phenylglycinol, s-2-tert-butoxycarbonylamino-2-phenylethanol, boc-phg-ol, n-boc-l-2-phenylglycinol, tert-butyl n-1s-2-hydroxy-1-phenylethyl carbamate, tert-butyl 1s-2-hydroxy-1-phenylethyl carbamate | |
| CC(C)(C)OC(=O)NC(CO)C1=CC=CC=C1 | |
| 1 g | |
| 7016462 | |
| ≥98.0% (GC) | |
| N-(tert-Butoxycarbonyl)-L-2-phenylglycinol |
Safety and Handling
TSCA : No