missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-L-2-phenylglycine 98.0+%, TCI America™

Catalog No. B32115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32115G 5 g
B321125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32115G Supplier TCI America Supplier No. B32115G
Only null left

Chemical Identifiers

CAS 2900-27-8
Molecular Formula C13H17NO4
Molecular Weight (g/mol) 251.282
MDL Number MFCD00065588
InChI Key HOBFSNNENNQQIU-JTQLQIEISA-N
Synonym boc-phg-oh, boc-l-phenylglycine, s-2-tert-butoxycarbonyl amino-2-phenylacetic acid, boc-l-alpha-phenylglycine, boc-l-phg-oh, boc-l-a-phenylglycine, n-boc-l-alpha-phenylglycine, n-boc-l-2-phenylglycine, s-boc-l-phenylglycine, n-boc-l-phenylglycine
PubChem CID 11010409
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-2-phenylacetic acid
SMILES CC(C)(C)OC(=O)NC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 93°C
Color White
Quantity 5 g
Formula Weight 251.28
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-L-2-phenylglycine