missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)iminodiacetic Acid 98.0+%, TCI America™
Supplier: TCI America B436825G
Specifications
| 126°C | |
| 56074-20-5 | |
| C9H15NO6 | |
| n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid | |
| CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O | |
| 25 g | |
| 2755971 | |
| ≥98.0% (HPLC,T) |
| N-(tert-Butoxycarbonyl)iminodiacetic Acid | |
| White | |
| MFCD00800351 | |
| QPGIJQUTGABQLQ-UHFFFAOYSA-N | |
| 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid | |
| 233.22 | |
| 233.22 | |
| Crystalline Powder |
Chemical Identifiers
| 56074-20-5 | |
| 233.22 | |
| QPGIJQUTGABQLQ-UHFFFAOYSA-N | |
| 2755971 | |
| CC(C)(C)OC(=O)N(CC(=O)O)CC(=O)O |
| C9H15NO6 | |
| MFCD00800351 | |
| n-boc-iminodiacetic acid, boc-dl-ida-oh, 2,2'-tert-butoxycarbonyl azanediyl diacetic acid, tert-butoxycarbonyl carboxymethyl amino acetic acid, n-tert-butoxycarbonyl iminodiacetic acid, glycine, n-carboxymethyl-n-1,1-dimethylethoxy carbonyl, 2-carboxymethyl-2-methylpropan-2-yl oxycarbonyl amino acetic acid, tert-butoxycarbonylimino bisacetic acid, n-t-butyloxycarbonyl iminodiacetic acid, n-tert-butoxycarbonyl-iminodiacetic acid | |
| 2-[carboxymethyl-[(2-methylpropan-2-yl)oxycarbonyl]amino]acetic acid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator