missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)glycine 98.0+%, TCI America™

Supplier:  TCI America B118525G

 View more versions of this product

Catalog No. B118525G


Only null left

Description

Description

Specifications

Specifications

89°C
4530-20-5
C7H13NO4
boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
CC(C)(C)OC(=O)NCC(O)=O
25 g
78288
≥98.0% (T)
N-(tert-Butoxycarbonyl)glycine
White
MFCD00002690
VRPJIFMKZZEXLR-UHFFFAOYSA-N
2-{[(tert-butoxy)carbonyl]amino}acetic acid
175.18
175.18
Crystalline Powder

Chemical Identifiers

4530-20-5
175.18
VRPJIFMKZZEXLR-UHFFFAOYSA-N
78288
CC(C)(C)OC(=O)NCC(O)=O
C7H13NO4
MFCD00002690
boc-glycine, boc-gly-oh, n-tert-butoxycarbonyl glycine, n-boc-glycine, n-boc-l-glycine, tert-butoxycarbonylglycine, t-butoxycarbonylglycine, 2-tert-butoxycarbonyl amino acetic acid, glycine, n-1,1-dimethylethoxy carbonyl, tert-butyloxycarbonylglycine
2-{[(tert-butoxy)carbonyl]amino}acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions