missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)glycine Methyl Ester 98.0+%, TCI America™

Catalog No. B39755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39755G 5 g
B397525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39755G Supplier TCI America Supplier No. B39755G

Chemical Identifiers

CAS 31954-27-5
Molecular Formula C8H15NO4
Molecular Weight (g/mol) 189.211
MDL Number MFCD00038267
InChI Key PHUZOEOLWIHIKH-UHFFFAOYSA-N
Synonym boc-glycine methyl ester, boc-gly-ome, n-tert-butoxycarbonyl glycine methyl ester, n-boc-glycine methyl ester, boc-glycinemethylester, methyl 2-tert-butoxycarbonyl amino acetate, methyl n-tert-butoxycarbonyl glycinate, glycine, n-1,1-dimethylethoxy carbonyl-, methyl ester, methyl tert-butoxycarbonyl amino acetate
PubChem CID 637609
IUPAC Name methyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
SMILES CC(C)(C)OC(=O)NCC(=O)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 189.21
Percent Purity ≥98.0% (N)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)glycine Methyl Ester