missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)ethanolamine, 98%

Catalog No. AC303170050
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303170050 5 mL Glass Bottle
AC303170250 25 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC303170050 Supplier Thermo Scientific Chemicals Supplier No. 303170050

CAS: 26690-80-2 | C7H15NO3 | 161.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 26690-80-2
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.2
MDL Number MFCD00056657
InChI Key GPTXCAZYUMDUMN-UHFFFAOYSA-N
Synonym n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
PubChem CID 2733206
IUPAC Name tert-butyl N-(2-hydroxyethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0420g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4485 to 1.4505
Linear Formula HOCH2CH2NHCO2C(CH3)3
Quantity 5 mL
Specific Gravity 1.042
Formula Weight 161.2
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)ethanolamine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent

RUO – Research Use Only