missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-DL-prolinol 98.0+%, TCI America™

Catalog No. B32735G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32735G 5 g
B32731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32735G Supplier TCI America Supplier No. B32735G

Chemical Identifiers

CAS 170491-63-1
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD01456556
InChI Key BFFLLBPMZCIGRM-UHFFFAOYSA-N
Synonym tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-2-hydroxymethyl pyrrolidine, n-boc-2-pyrrolidinemethanol, boc-dl-prolinol, 2-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, tert-butyl 2-hydroxymethyl-1-pyrrolidinecarboxylate, 2-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester, 1-pyrrolidinecarboxylic acid, 2-hydroxymethyl-, 1,1-dimethylethyl ester, r-1-boc-2-hydroxymethyl pyrrolidine, n-boc-dl-prolinol
PubChem CID 550865
IUPAC Name tert-butyl 2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 54°C
Color White
Quantity 5 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-DL-prolinol