missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-DL-phenylalaninol 96.0+%, TCI America™

Catalog No. b32691g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32691G 1 g
B32695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B32691G Supplier TCI America Supplier No. B32691G

Chemical Identifiers

CAS 145149-48-0
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.326
MDL Number MFCD01457313
InChI Key LDKDMDVMMCXTMO-UHFFFAOYSA-N
Synonym N-Boc-DL-phenylalaninol, 2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol
PubChem CID 545866
IUPAC Name tert-butyl N-(1-hydroxy-3-phenylpropan-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CO

Specifications

Melting Point 86°C
Color White-Yellow
Quantity 1 g
Formula Weight 251.33
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-DL-phenylalaninol