missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(tert-Butoxycarbonyl)-D-tert-leucine, TCI America™

$44.31 - $146.67

Chemical Identifiers

CAS 124655-17-0
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.292
MDL Number MFCD00065575
InChI Key LRFZIPCTFBPFLX-ZETCQYMHSA-N
Synonym boc-d-tert-leucine, n-boc-d-tert-leucine, boc-d-tle-oh, r-n-tert-butoxycarbonyl-tert-leucine, r-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-d-alpha-t-butylglycine, boc-tbu-d-gly-oh, boc-d-t-leu, d-valine, n-1,1-dimethylethoxy carbonyl-3-methyl, n-tert-butoxycarbonyl-d-tert-leucine
PubChem CID 7005057
IUPAC Name (2R)-3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B39951G View Documents TCI America
B39951G
1g Each for $44.31
 
B39955G View Documents TCI America
B39955G
5g Each for $146.67
 
Specifications

Chemical Identifiers

124655-17-0
231.292
LRFZIPCTFBPFLX-ZETCQYMHSA-N
7005057
CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
C11H21NO4
MFCD00065575
boc-d-tert-leucine, n-boc-d-tert-leucine, boc-d-tle-oh, r-n-tert-butoxycarbonyl-tert-leucine, r-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-d-alpha-t-butylglycine, boc-tbu-d-gly-oh, boc-d-t-leu, d-valine, n-1,1-dimethylethoxy carbonyl-3-methyl, n-tert-butoxycarbonyl-d-tert-leucine
(2R)-3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

White
1g
C11H21NO4
boc-d-tert-leucine, n-boc-d-tert-leucine, boc-d-tle-oh, r-n-tert-butoxycarbonyl-tert-leucine, r-2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-d-alpha-t-butylglycine, boc-tbu-d-gly-oh, boc-d-t-leu, d-valine, n-1,1-dimethylethoxy carbonyl-3-methyl, n-tert-butoxycarbonyl-d-tert-leucine
CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
231.292
231.29
≥98.0% (HPLC,T)
120°C
124655-17-0
MFCD00065575
LRFZIPCTFBPFLX-ZETCQYMHSA-N
(2R)-3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
7005057
Crystal-Powder at 20°C
N-(tert-Butoxycarbonyl)-D-tert-leucine
Safety and Handling

Safety and Handling

TSCA : No