missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-prolinol 98.0+%, TCI America™

Catalog No. B30761G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30761G 1 g
B30765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30761G Supplier TCI America Supplier No. B30761G
Only null left

Chemical Identifiers

CAS 83435-58-9
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.266
MDL Number MFCD00040580
InChI Key BFFLLBPMZCIGRM-MRVPVSSYSA-N
Synonym boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate
PubChem CID 688279
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

Melting Point 62°C
Color White-Yellow
Quantity 1 g
Formula Weight 201.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-prolinol