missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-tert-Butoxycarbonyl-D-prolinol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 344550010
| Quantity | 1g |
|---|---|
| Packaging | Glass bottle |
Description
Chemical Identifiers
| 83435-58-9 | |
| 201.266 | |
| BFFLLBPMZCIGRM-MRVPVSSYSA-N | |
| 688279 | |
| CC(C)(C)OC(=O)N1CCCC1CO |
| C10H19NO3 | |
| MFCD00040580 | |
| boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate | |
| tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate |
Specifications
| N-tert-Butoxycarbonyl-D-prolinol | |
| 83435-58-9 | |
| 100.0 | |
| 97.5% min. (GC) | |
| C10H19NO3 | |
| MFCD00040580 | |
| boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate | |
| BFFLLBPMZCIGRM-MRVPVSSYSA-N | |
| tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate | |
| 688279 | |
| 98% |
| 98% | |
| 97.5 | |
| Authentic | |
| Glass bottle | |
| 1g | |
| + 49.00 | |
| Solubility in water: insoluble in water. | |
| CC(C)(C)OC(=O)N1CCCC1CO | |
| 201.266 | |
| 201.27 |