missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-D-proline 98.0+%, TCI America™
$61.51 - $126.76
Chemical Identifiers
| CAS | 37784-17-1 |
|---|---|
| Molecular Formula | C10H17NO4 |
| Molecular Weight (g/mol) | 215.249 |
| MDL Number | MFCD00063226 |
| InChI Key | ZQEBQGAAWMOMAI-SSDOTTSWSA-N |
| Synonym | n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-proline |
| PubChem CID | 688022 |
| IUPAC Name | (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
| SMILES | CC(C)(C)OC(=O)N1CCCC1C(=O)O |
Chemical Identifiers
| 37784-17-1 | |
| 215.249 | |
| ZQEBQGAAWMOMAI-SSDOTTSWSA-N | |
| 688022 | |
| CC(C)(C)OC(=O)N1CCCC1C(=O)O |
| C10H17NO4 | |
| MFCD00063226 | |
| n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-proline | |
| (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid |
Specifications
| 37784-17-1 | |
| White | |
| MFCD00063226 | |
| n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-proline | |
| CC(C)(C)OC(=O)N1CCCC1C(=O)O | |
| 215.249 | |
| 215.25 | |
| Crystalline Powder |
| 135°C | |
| C10H17NO4 | |
| 5 g | |
| ZQEBQGAAWMOMAI-SSDOTTSWSA-N | |
| (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid | |
| 688022 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-D-proline |
Safety and Handling
TSCA : No