missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-proline 98.0+%, TCI America™

Catalog No. B29775G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29775G 5 g
B297725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29775G Supplier TCI America Supplier No. B29775G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 37784-17-1
Molecular Formula C10H17NO4
Molecular Weight (g/mol) 215.249
MDL Number MFCD00063226
InChI Key ZQEBQGAAWMOMAI-SSDOTTSWSA-N
Synonym n-boc-d-proline, boc-d-proline, boc-d-pro-oh, r-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, n-tert-butoxycarbonyl-d-proline, 2r-1-tert-butoxy carbonyl pyrrolidine-2-carboxylic acid, n-t-boc-d-proline, boc-d-pro, n-alpha-t-boc-d-proline, 1-tert-butoxycarbonyl-d-proline
PubChem CID 688022
IUPAC Name (2R)-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1C(=O)O

Specifications

Melting Point 135°C
Color White
Quantity 5 g
Formula Weight 215.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-proline