missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-phenylalaninol 97.0+%, TCI America™

Catalog No. B19661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19661G 1 g
B19665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19661G Supplier TCI America Supplier No. B19661G

Chemical Identifiers

CAS 106454-69-7
Molecular Formula C14H21NO3
Molecular Weight (g/mol) 251.33
MDL Number MFCD00216472
InChI Key LDKDMDVMMCXTMO-GFCCVEGCSA-N
Synonym n-boc-d-phenylalaninol, boc-d-phenylalaninol, r-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate, n-tert-butoxycarbonyl-d-phenylalaninol, r-2-boc-amino-3-phenyl-1-propanol, r-+-2-tert-butoxycarbonylamino-3-phenyl-1-propanol, r-2-tert-butoxycarbonylamino-3-phenyl-1-propanol, tert-butyl n-2r-1-hydroxy-3-phenylpropan-2-yl carbamate, pubchem5739, r-n-tert-butoxycarbonyl-beta-phenylalaninol
PubChem CID 7019371
IUPAC Name tert-butyl N-[(2R)-1-hydroxy-3-phenylpropan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 95°C
Color White
Quantity 1 g
Formula Weight 251.33
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-phenylalaninol