missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-leucine Hydrate 98.0+%, TCI America™

Catalog No. B30075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30075G 5 g
B300725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30075G Supplier TCI America Supplier No. B30075G

Chemical Identifiers

CAS 16937-99-8
Molecular Formula C11H23NO5
Molecular Weight (g/mol) 249.307
MDL Number MFCD00038294
InChI Key URQQEIOTRWJXBA-DDWIOCJRSA-N
Synonym boc-d-leucine monohydrate, r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leu-oh.h2o, n-boc-d-leucine monohydrate, boc-d-leu-oh h2o, d-leucine, n-1,1-dimethylethoxy carbonyl-, monohydrate, 2r-2-tert-butoxycarbonyl amino-4-methylpentanoic acid hydrate, boc-d-leucine h2o, boc-d-leucine hydrate, n-boc-d-leucinemonohydrate
PubChem CID 2724763
IUPAC Name (2R)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate
SMILES CC(C)CC(C(=O)O)NC(=O)OC(C)(C)C.O

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-leucine Hydrate