missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-D-glutamic Acid 97.0+%, TCI America™
$93.21 - $277.75
Chemical Identifiers
| CAS | 34404-28-9 |
|---|---|
| Molecular Formula | C10H17NO6 |
| Molecular Weight (g/mol) | 247.247 |
| MDL Number | MFCD00190790 |
| InChI Key | AQTUACKQXJNHFQ-ZCFIWIBFSA-N |
| Synonym | boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid |
| PubChem CID | 7019046 |
| IUPAC Name | (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid |
| SMILES | CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O |
Chemical Identifiers
| 34404-28-9 | |
| 247.247 | |
| AQTUACKQXJNHFQ-ZCFIWIBFSA-N | |
| 7019046 | |
| CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O |
| C10H17NO6 | |
| MFCD00190790 | |
| boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid | |
| (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid |
Specifications
| 34404-28-9 | |
| White-Yellow | |
| MFCD00190790 | |
| boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid | |
| CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O | |
| 247.247 | |
| 247.25 | |
| Crystalline Powder |
| 108°C | |
| C10H17NO6 | |
| 5 g | |
| AQTUACKQXJNHFQ-ZCFIWIBFSA-N | |
| (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid | |
| 7019046 | |
| ≥97.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-D-glutamic Acid |
Safety and Handling
TSCA : No