missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-D-glutamic Acid 97.0+%, TCI America™

Catalog No. B29875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29875G 5 g
B298725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29875G Supplier TCI America Supplier No. B29875G
Only null left

Chemical Identifiers

CAS 34404-28-9
Molecular Formula C10H17NO6
Molecular Weight (g/mol) 247.247
MDL Number MFCD00190790
InChI Key AQTUACKQXJNHFQ-ZCFIWIBFSA-N
Synonym boc-d-glu-oh, boc-d-glutamic acid, n-tert-butoxycarbonyl-d-glutamic acid, r-2-tert-butoxycarbonyl amino pentanedioic acid, d-glutamic acid, n-1,1-dimethylethoxy carbonyl, 2r-2-tert-butoxycarbonyl amino pentanedioic acid, pubchem20973, n-boc-d-glutamic acid, ksc493m2t, 2r-2-2-methylpropan-2-yl oxycarbonylamino pentanedioic acid
PubChem CID 7019046
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanedioic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)O)C(=O)O

Specifications

Melting Point 108°C
Color White-Yellow
Quantity 5 g
Formula Weight 247.25
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-D-glutamic Acid