missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 91°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 251.28 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-(tert-Butoxycarbonyl)-D-2-phenylglycine |
Safety and Handling
| TSCA | No |