missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-cis-4-hydroxy-L-proline Methyl Ester 97.0+%, TCI America™

Catalog No. p-7123286
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39871G 1 g
B39875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39871G Supplier TCI America Supplier No. B39871G
Only null left

Chemical Identifiers

CAS 102195-79-9
Molecular Formula C11H19NO5
Molecular Weight (g/mol) 245.275
MDL Number MFCD00237541
InChI Key MZMNEDXVUJLQAF-YUMQZZPRSA-N
Synonym n-boc-cis-4-hydroxy-l-proline methyl ester, boc-cis-hyp-ome, 2s,4s-1-tert-butyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, cis-boc-hyp-ome, boc-cis-4-hydroxy-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4s-4-hydroxypyrrolidine-1,2-dicarboxylate, methyl cis-1-boc-4-hydroxy-l-prolinate, 2s,4s-4-hydroxy-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester 2-methyl ester, methyl 4s-n-boc-4-hydroxy-l-prolinate, o1-tert-butyl o2-methyl 2s,4s-4-hydroxypyrrolidine-1,2-dicarboxylate
PubChem CID 10752873
IUPAC Name 1-O-tert-butyl 2-O-methyl (2S,4S)-4-hydroxypyrrolidine-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CC(CC1C(=O)OC)O

Specifications

Melting Point 82°C
Color White
Quantity 1 g
Formula Weight 245.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-cis-4-hydroxy-L-proline Methyl Ester