missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-7-aminoheptanoic Acid 98.0+%, TCI America™

Catalog No. B42921G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42921G 1 g
B42925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42921G Supplier TCI America Supplier No. B42921G
Only null left

Chemical Identifiers

CAS 60142-89-4
Molecular Formula C12H23NO4
Molecular Weight (g/mol) 245.319
MDL Number MFCD00063356
InChI Key HJENAZQPOGVAEK-UHFFFAOYSA-N
Synonym N-Boc-7-aminoheptanoic Acid, N-(tert-Butoxycarbonyl)-7-aminoenanthic Acid, N-Boc-7-aminoenanthic Acid, 7-(tert-Butoxycarbonylamino)heptanoic Acid, 7-(Boc-amino)heptanoic Acid, 7-(tert-Butoxycarbonylamino)enanthic Acid, 7-(Boc-amino)enanthic Acid, Boc-7-Ah
PubChem CID 546122
IUPAC Name 7-[(2-methylpropan-2-yl)oxycarbonylamino]heptanoic acid
SMILES CC(C)(C)OC(=O)NCCCCCCC(=O)O

Specifications

Melting Point 57°C
Color White
Quantity 1 g
Formula Weight 245.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-7-aminoheptanoic Acid
TSCA No
Recommended Storage Refrigerator