missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-5-aminovaleric Acid 98.0+%, TCI America™

Catalog No. B28955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28955G 5 g
B289525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28955G Supplier TCI America Supplier No. B28955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 27219-07-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD00076903
InChI Key GFMRZAMDGJIWRB-UHFFFAOYSA-N
Synonym boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid
PubChem CID 545848
IUPAC Name 5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NCCCCC(=O)O

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 168°C
Quantity 5 g
Formula Weight 217.27
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-5-aminovaleric Acid