missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-5-aminovaleric Acid 98.0+%, TCI America™
Supplier: TCI America B289525G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 27219-07-4 | |
| 217.265 | |
| GFMRZAMDGJIWRB-UHFFFAOYSA-N | |
| 545848 | |
| CC(C)(C)OC(=O)NCCCCC(=O)O |
| C10H19NO4 | |
| MFCD00076903 | |
| boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid | |
| 5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid |
Specifications
| N-(tert-Butoxycarbonyl)-5-aminovaleric Acid | |
| 52°C | |
| 168°C | |
| 25 g | |
| boc-5-ava-oh, 5-tert-butoxycarbonylamino valeric acid, boc-5-aminovaleric acid, 5-boc-amino valeric acid, 5-tert-butoxycarbonyl amino pentanoic acid, 5-boc-amino-pentanoic acid, n-boc-5-aminopentanoic acid, pentanoic acid, 5-1,1-dimethylethoxy carbonyl amino, n-boc-5-aminovaleric acid | |
| CC(C)(C)OC(=O)NCCCCC(=O)O | |
| 217.265 | |
| 217.27 | |
| Crystalline Powder |
| 27219-07-4 | |
| White-Yellow | |
| C10H19NO4 | |
| MFCD00076903 | |
| GFMRZAMDGJIWRB-UHFFFAOYSA-N | |
| 5-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid | |
| 545848 | |
| ≥98.0% (GC,T) |
Safety and Handling
TSCA : No