missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine 98.0+%, TCI America™

Catalog No. B42891G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42891G Supplier TCI America Supplier No. B42891G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147923-08-8
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.407
MDL Number MFCD00191184
InChI Key NBVVKAUSAGHTSU-KRWDZBQOSA-N
Synonym boc-l-4,4'-biphenylalanine, boc-4-phenyl-phe-oh, boc-3-4-biphenylyl-l-alanine, boc-bip-oh, boc-4-phenyl-l-phenylalanine, s-3-1,1'-biphenyl-4-yl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-boc-l-biphenylalanine, boc-4-biphenyl-l-ala, boc-l-4,4'-biphenylphenylalanine, n-boc-4-phenyl-l-phenylalanine
PubChem CID 2761796
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylphenyl)propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C2=CC=CC=C2)C(=O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 341.41
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-phenyl-L-phenylalanine