missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine 98.0+%, TCI America™

Catalog No. B33045G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33045G 5 g
B33041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33045G Supplier TCI America Supplier No. B33045G

Chemical Identifiers

CAS 41153-30-4
Molecular Formula C14H18FNO4
Molecular Weight (g/mol) 283.299
MDL Number MFCD00079672
InChI Key RCXSXRAUMLKRRL-NSHDSACASA-N
Synonym boc-l-4-fluorophenylalanine, boc-phe 4-f-oh, boc-l-4-fluorophe, boc-4-fluoro-l-phenylalanine, boc-4-fluoro-l-phe-oh, n-tert-butoxycarbonyl-4-fluoro-l-phenylalanine, s-n-boc-4-fluorophenylalanine, boc-l-phe 4-f-oh, boc-p-fluoro-l-phenylalanine, boc-p-fluoro-phe-oh
PubChem CID 7020910
IUPAC Name (2S)-3-(4-fluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)F)C(=O)O

Specifications

Melting Point 85°C
Color White
Quantity 5 g
Formula Weight 283.30
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine