missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine 98.0+%, TCI America™
$96.94 - $467.87
Chemical Identifiers
| CAS | 131724-45-3 |
|---|---|
| Molecular Formula | C15H18N2O4 |
| Molecular Weight (g/mol) | 290.319 |
| MDL Number | MFCD00672526 |
| InChI Key | RMBLTLXJGNILPG-LBPRGKRZSA-N |
| Synonym | boc-phe 4-cn-oh, boc-l-4-cyanophenylalanine, boc-4-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, boc-d-4-cyanophenylalanine, n-boc-4-cyanophenylalanine, s-n-boc-4-cyanophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2s-3-4-cyanophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-l-4-cn-phe-oh |
| PubChem CID | 7020841 |
| IUPAC Name | (2S)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C#N)C(=O)O |
Chemical Identifiers
| 131724-45-3 | |
| 290.319 | |
| RMBLTLXJGNILPG-LBPRGKRZSA-N | |
| 7020841 | |
| CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C#N)C(=O)O |
| C15H18N2O4 | |
| MFCD00672526 | |
| boc-phe 4-cn-oh, boc-l-4-cyanophenylalanine, boc-4-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, boc-d-4-cyanophenylalanine, n-boc-4-cyanophenylalanine, s-n-boc-4-cyanophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2s-3-4-cyanophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-l-4-cn-phe-oh | |
| (2S)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 131724-45-3 | |
| White | |
| MFCD00672526 | |
| boc-phe 4-cn-oh, boc-l-4-cyanophenylalanine, boc-4-cyano-l-phenylalanine, s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, boc-d-4-cyanophenylalanine, n-boc-4-cyanophenylalanine, s-n-boc-4-cyanophenylalanine, 2s-2-tert-butoxycarbonyl amino-3-4-cyanophenyl propanoic acid, 2s-3-4-cyanophenyl-2-2-methylpropan-2-yl oxycarbonylamino propanoic acid, boc-l-4-cn-phe-oh | |
| CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)C#N)C(=O)O | |
| 290.319 | |
| 290.32 | |
| Crystalline Powder |
| 147°C | |
| C15H18N2O4 | |
| 1 g | |
| RMBLTLXJGNILPG-LBPRGKRZSA-N | |
| (2S)-3-(4-cyanophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 7020841 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine |
Safety and Handling
TSCA : No