missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine 98.0+%, TCI America™

Catalog No. B36131G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36131G 1 g
B36135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36131G Supplier TCI America Supplier No. B36131G

Chemical Identifiers

CAS 57292-44-1
Molecular Formula C14H18ClNO4
Molecular Weight (g/mol) 299.75
MDL Number MFCD00076978
InChI Key BETBOAZCLSJOBQ-LLVKDONJSA-N
Synonym boc-d-phe 4-cl-oh, boc-4-chloro-d-phenylalanine, boc-d-4-chlorophe, boc-d-4-chlorophenylalanine, n-tert-butoxycarbonyl-4-chloro-d-phenylalanine, n-boc-4-chloro-d-phenylalanine, r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, r-n-boc-4-chlorophenylalanine, 2r-2-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, d-phenylalanine, 4-chloro-n-1,1-dimethylethoxy carbonyl
PubChem CID 7018791
IUPAC Name (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-chlorophenyl)propanoic acid
SMILES CC(C)(C)OC(=O)N[C@H](CC1=CC=C(Cl)C=C1)C(O)=O

Specifications

Melting Point 110°C
Color White
Quantity 1 g
Formula Weight 299.75
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-4-chloro-D-phenylalanine