missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-4-aminobutyric Acid 98.0+%, TCI America™
$72.70 - $337.40
Chemical Identifiers
| CAS | 57294-38-9 |
|---|---|
| Molecular Formula | C9H17NO4 |
| Molecular Weight (g/mol) | 203.24 |
| MDL Number | MFCD00037313 |
| InChI Key | HIDJWBGOQFTDLU-UHFFFAOYSA-N |
| Synonym | boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid |
| PubChem CID | 294894 |
| IUPAC Name | 4-{[(tert-butoxy)carbonyl]amino}butanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC(O)=O |
Chemical Identifiers
| 57294-38-9 | |
| 203.24 | |
| HIDJWBGOQFTDLU-UHFFFAOYSA-N | |
| 294894 | |
| CC(C)(C)OC(=O)NCCCC(O)=O |
| C9H17NO4 | |
| MFCD00037313 | |
| boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid | |
| 4-{[(tert-butoxy)carbonyl]amino}butanoic acid |
Specifications
| 58°C | |
| 82°C | |
| C9H17NO4 | |
| boc-gaba-oh, n-boc-gamma-aminobutyric acid, boc-gamma-abu-oh, 4-tert-butoxycarbonyl amino butanoic acid, 4-boc-amino butyric acid, 4-tert-butoxycarbonylaminobutyric acid, 4-tert-butoxycarbonylamino butyric acid, boc-gaba, 4-tert-butoxycarbonylamino butanoic acid | |
| CC(C)(C)OC(=O)NCCCC(O)=O | |
| 5 g | |
| 294894 | |
| ≥98.0% (T) | |
| N-(tert-Butoxycarbonyl)-4-aminobutyric Acid |
| 57294-38-9 | |
| White-Yellow | |
| MFCD00037313 | |
| HIDJWBGOQFTDLU-UHFFFAOYSA-N | |
| 4-{[(tert-butoxy)carbonyl]amino}butanoic acid | |
| 203.24 | |
| 203.24 | |
| Crystalline Powder |
Safety and Handling
TSCA : No