missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine 95.0+%, TCI America™
$367.41 - $367.41
Chemical Identifiers
| CAS | 205445-51-8 |
|---|---|
| Molecular Formula | C14H17F2NO4 |
| Molecular Weight (g/mol) | 301.29 |
| MDL Number | MFCD00672519 |
| InChI Key | CYAOPVHXASZUDE-LLVKDONJSA-N |
| Synonym | boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected |
| PubChem CID | 2736194 |
| IUPAC Name | (2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O |
Chemical Identifiers
| 205445-51-8 | |
| 301.29 | |
| CYAOPVHXASZUDE-LLVKDONJSA-N | |
| 2736194 | |
| CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O |
| C14H17F2NO4 | |
| MFCD00672519 | |
| boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected | |
| (2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
Specifications
| 205445-51-8 | |
| C14H17F2NO4 | |
| 5 g | |
| CYAOPVHXASZUDE-LLVKDONJSA-N | |
| (2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
| 2736194 | |
| ≥95.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine |
| White | |
| MFCD00672519 | |
| boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected | |
| CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O | |
| 301.29 | |
| 301.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : No