missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine 95.0+%, TCI America™

Catalog No. B36265G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36265G Supplier TCI America Supplier No. B36265G
Only null left

Chemical Identifiers

CAS 205445-51-8
Molecular Formula C14H17F2NO4
Molecular Weight (g/mol) 301.29
MDL Number MFCD00672519
InChI Key CYAOPVHXASZUDE-LLVKDONJSA-N
Synonym boc-d-3,4-difluorophenylalanine, boc-3,4-difluoro-d-phenylalanine, boc-d-phe 3,4-dif-oh, boc-d-3,4-difluorophe, boc-d-phe 3,4-f2-oh, n-tert-butoxycarbonyl-3,4-difluoro-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3-3,4-difluorophenyl propanoic acid, n-boc-3,4-difluoro-d-phenylalanine, n-alpha-t-butyloxycarbonyl-d-3,4-difluorophenylalanine, 3,4-difluoro-d-phenylalanine, boc protected
PubChem CID 2736194
IUPAC Name (2R)-3-(3,4-difluorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC(=C(C=C1)F)F)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 301.29
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3,4-difluoro-D-phenylalanine