missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine 98.0+%, TCI America™

$282.90 - $282.90

Chemical Identifiers

CAS 58438-04-3
Molecular Formula C18H21NO4
Molecular Weight (g/mol) 315.369
MDL Number MFCD00079671
InChI Key URKWHOVNPHQQTM-HNNXBMFYSA-N
Synonym boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
PubChem CID 7020908
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B36171G
View Documents
TCI America
B36171G
1 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

58438-04-3
315.369
URKWHOVNPHQQTM-HNNXBMFYSA-N
7020908
CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O
C18H21NO4
MFCD00079671
boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid

Specifications

100°C
White
MFCD00079671
URKWHOVNPHQQTM-HNNXBMFYSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
315.369
315.37
Crystalline Powder
58438-04-3
C18H21NO4
boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O
1 g
7020908
≥98.0% (HPLC,T)
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents