missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine 98.0+%, TCI America™

Catalog No. B36171G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36171G Supplier TCI America Supplier No. B36171G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 58438-04-3 | Formula: C18H21NO4 | Weight: 315.369 g/mol

Chemical Identifiers

CAS 58438-04-3
Molecular Formula C18H21NO4
Molecular Weight (g/mol) 315.369
MDL Number MFCD00079671
InChI Key URKWHOVNPHQQTM-HNNXBMFYSA-N
Synonym boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
PubChem CID 7020908
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O

Specifications

Melting Point 100°C
Color White
Quantity 1 g
Formula Weight 315.37
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine