missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine 98.0+%, TCI America™
$96.94 - $374.68
Chemical Identifiers
| CAS | 76985-10-9 |
|---|---|
| Molecular Formula | C18H21NO4 |
| Molecular Weight (g/mol) | 315.37 |
| MDL Number | MFCD00076900 |
| InChI Key | URKWHOVNPHQQTM-OAHLLOKOSA-N |
| Synonym | boc-d-2-nal-oh, boc-3-2-naphthyl-d-alanine, n-tert-butoxycarbonyl-3-2-naphthyl-d-alanine, boc-2-d-nal-oh, r-n-boc-2-naphthylalanine, boc-d-3-2-naphthyl-alanine, n-boc-3-2-naphthyl-d-alanine, n-boc-2-2-naphthyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid |
| PubChem CID | 2734481 |
| IUPAC Name | (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoic acid |
| SMILES | CC(C)(C)OC(=O)N[C@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O |
Chemical Identifiers
| 76985-10-9 | |
| 315.37 | |
| URKWHOVNPHQQTM-OAHLLOKOSA-N | |
| 2734481 | |
| CC(C)(C)OC(=O)N[C@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O |
| C18H21NO4 | |
| MFCD00076900 | |
| boc-d-2-nal-oh, boc-3-2-naphthyl-d-alanine, n-tert-butoxycarbonyl-3-2-naphthyl-d-alanine, boc-2-d-nal-oh, r-n-boc-2-naphthylalanine, boc-d-3-2-naphthyl-alanine, n-boc-3-2-naphthyl-d-alanine, n-boc-2-2-naphthyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid | |
| (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoic acid |
Specifications
| 95°C | |
| White | |
| MFCD00076900 | |
| URKWHOVNPHQQTM-OAHLLOKOSA-N | |
| (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoic acid | |
| 315.37 | |
| 315.37 | |
| Crystalline Powder |
| 76985-10-9 | |
| C18H21NO4 | |
| boc-d-2-nal-oh, boc-3-2-naphthyl-d-alanine, n-tert-butoxycarbonyl-3-2-naphthyl-d-alanine, boc-2-d-nal-oh, r-n-boc-2-naphthylalanine, boc-d-3-2-naphthyl-alanine, n-boc-3-2-naphthyl-d-alanine, n-boc-2-2-naphthyl-d-alanine, r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid, 2r-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid | |
| CC(C)(C)OC(=O)N[C@H](CC1=CC2=CC=CC=C2C=C1)C(O)=O | |
| 1 g | |
| 2734481 | |
| ≥98.0% (HPLC,T) | |
| N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine |
Safety and Handling
TSCA : No