missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-2-methyl-1,3-diaminopropane 98.0+%, TCI America™

Catalog No. p-7119774
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28101G 1 g
B28105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28101G Supplier TCI America Supplier No. B28101G

Chemical Identifiers

CAS 480452-05-9
Molecular Formula C9H20N2O2
Molecular Weight (g/mol) 188.271
InChI Key ORDMEFMUSKDHOL-UHFFFAOYSA-N
Synonym N-(3-Amino-2-methylpropyl)carbamic Acid tert-Butyl Ester, N-Boc-2-methyl-1,3-diaminopropane, N-Boc-2-methyl-1,3-propanediamine, N-(tert-Butoxycarbonyl)-2-methyl-1,3-propanediamine, tert-Butyl N-(3-Amino-2-methylpropyl)carbamate
PubChem CID 44629801
IUPAC Name tert-butyl N-(3-amino-2-methylpropyl)carbamate
SMILES CC(CN)CNC(=O)OC(C)(C)C

Specifications

Color Colorless
Quantity 1 g
Formula Weight 188.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-2-methyl-1,3-diaminopropane