missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-1,4-diaminobutane 98.0+%, TCI America™
$110.11 - $1,036.64
Chemical Identifiers
| CAS | 68076-36-8 |
|---|---|
| Molecular Formula | C9H20N2O2 |
| Molecular Weight (g/mol) | 188.271 |
| MDL Number | MFCD00210019 |
| InChI Key | ZFQWJXFJJZUVPI-UHFFFAOYSA-N |
| Synonym | tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane |
| PubChem CID | 4351 |
| IUPAC Name | tert-butyl N-(4-aminobutyl)carbamate |
| SMILES | CC(C)(C)OC(=O)NCCCCN |
Chemical Identifiers
| 68076-36-8 | |
| 188.271 | |
| ZFQWJXFJJZUVPI-UHFFFAOYSA-N | |
| 4351 | |
| CC(C)(C)OC(=O)NCCCCN |
| C9H20N2O2 | |
| MFCD00210019 | |
| tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane | |
| tert-butyl N-(4-aminobutyl)carbamate |
Specifications
| 68076-36-8 | |
| C9H20N2O2 | |
| tert-butyl n-4-aminobutyl carbamate, n-boc-1,4-butanediamine, n-boc-1,4-diaminobutane, tert-butyl 4-aminobutyl carbamate, tert-butyl 4-aminobutylcarbamate, boc-1,4-diaminobutane, n-boc-1,4-butandiamine, n-4-aminobutyl carbamic acid tert-butyl ester, n-tert-butoxycarbonyl-1,4-diaminobutane, mono-4-n-boc-1,4-diaminobutane | |
| CC(C)(C)OC(=O)NCCCCN | |
| 188.271 | |
| 188.27 | |
| Liquid |
| Yellow | |
| MFCD00210019 | |
| ZFQWJXFJJZUVPI-UHFFFAOYSA-N | |
| tert-butyl N-(4-aminobutyl)carbamate | |
| 4351 | |
| ≥98.0% (GC,T) | |
| N-(tert-Butoxycarbonyl)-1,4-diaminobutane |
Safety and Handling
TSCA : No