missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-1,2-diaminoethane 97.0+%, TCI America™
Supplier: TCI America A137125G
Specifications
| N-(tert-Butoxycarbonyl)-1,2-diaminoethane | |
| 105°C | |
| C7H16N2O2 | |
| 2735 | |
| AOCSUUGBCMTKJH-UHFFFAOYSA-N | |
| tert-butyl N-(2-aminoethyl)carbamate | |
| 160.22 | |
| 160.22 | |
| Liquid |
| 57260-73-8 | |
| Yellow | |
| MFCD00191871 | |
| n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane | |
| CC(C)(C)OC(=O)NCCN | |
| 25 g | |
| 187201 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 57260-73-8 | |
| 160.22 | |
| AOCSUUGBCMTKJH-UHFFFAOYSA-N | |
| 187201 | |
| CC(C)(C)OC(=O)NCCN |
| C7H16N2O2 | |
| MFCD00191871 | |
| n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane | |
| tert-butyl N-(2-aminoethyl)carbamate |
Safety and Handling
TSCA : No