missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)-1,2-diaminoethane 97.0+%, TCI America™

Catalog No. A13711G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13711G 1 g
A13715G 5 g
A137125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A13711G Supplier TCI America Supplier No. A13711G

Chemical Identifiers

CAS 57260-73-8
Molecular Formula C7H16N2O2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00191871
InChI Key AOCSUUGBCMTKJH-UHFFFAOYSA-N
Synonym n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane
PubChem CID 187201
IUPAC Name tert-butyl N-(2-aminoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCN

Specifications

Boiling Point 105°C
Color Yellow
UN Number 2735
Quantity 1 g
Formula Weight 160.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)-1,2-diaminoethane