missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl D-Biotinate 97.0+%, TCI America™
Supplier: TCI America S04911G
Specifications
| N-Succinimidyl D-Biotinate | |
| 140°C | |
| C14H19N3O5S | |
| 1 g | |
| YMXHPSHLTSZXKH-RVBZMBCESA-N | |
| (2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate | |
| 6710714 | |
| ≥97.0% (HPLC,N) |
| 35013-72-0 | |
| White-Yellow | |
| MFCD00078531 | |
| +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester | |
| C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3 | |
| 341.382 | |
| 341.38 | |
| Crystalline Powder |
Chemical Identifiers
| 35013-72-0 | |
| 341.382 | |
| YMXHPSHLTSZXKH-RVBZMBCESA-N | |
| 6710714 | |
| C1CC(=O)N(C1=O)OC(=O)CCCCC2C3C(CS2)NC(=O)N3 |
| C14H19N3O5S | |
| MFCD00078531 | |
| +-biotin n-hydroxysuccinimide ester, biotin-nhs, biotin nhs, 2,5-dioxopyrrolidin-1-yl 5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanoate, n-hydroxysuccinimidobiotin, d-biotinyl-osu, biotin-osu, n-succinimidyl d-biotinate, bhse, biotin n-hydroxysuccinimide ester | |
| (2,5-dioxopyrrolidin-1-yl) 5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoate |
Safety and Handling
TSCA : No
Recommended Storage : Freezer